Product Details

SNP ID
rs7387694
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119885261 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCAAAAAGTGGGGTTTATTCCC[G/T]GTAGTGGTATATGACTGTGTTTTAC
Phenotype
MIM: 612974
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DEPTOR PubMed Links
Additional Information
For this assay, SNP(s) [rs371094002] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DEPTOR
Gene Name
DEP domain containing MTOR-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283012.1 Intron NP_001269941.1
NM_022783.3 Intron NP_073620.2
XM_017013735.1 Intron XP_016869224.1

View Full Product Details