Product Details
- SNP ID
-
rs6795223
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51829910 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCGTGCGCACAGCCACCTGATAG[G/T]TGGGGCTCAGCACAGGAAGTGGAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IQCF3
PubMed Links
Gene Details
- Gene
- IQCF3
- Gene Name
- IQ motif containing F3
View Full Product Details