Product Details

SNP ID
rs7951221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16787526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTAATGATTCACAAAGAAAACA[C/T]TCATTTCCTTACACGCTCACAAATG
Phenotype
MIM: 612686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC283278 PubMed Links

Gene Details

Gene
LOC283278
Gene Name
pleckstrin homology domain-containing family A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017018657.1 Intron XP_016874146.1
XM_017018658.1 Intron XP_016874147.1
Gene
PLEKHA7
Gene Name
pleckstrin homology domain containing A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175058.4 Intron NP_778228.3
XM_017017240.1 Intron XP_016872729.1
XM_017017241.1 Intron XP_016872730.1
XM_017017242.1 Intron XP_016872731.1
XM_017017243.1 Intron XP_016872732.1

View Full Product Details