Product Details

SNP ID
rs7951676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16747703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTCATTTGCTTTCTCTCCATGA[G/T]ATATACTTGAGGCAGCAGAAAGAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf58 PubMed Links

Gene Details

Gene
C11orf58
Gene Name
chromosome 11 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014267.5 Intron NP_055082.1
XM_017017143.1 Intron XP_016872632.1

View Full Product Details