Product Details

SNP ID
rs6864377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:62413486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTCAGCGACTCCTGTAGATTT[A/G]TATTTGGGGGTATTTGTGATTCAGC
Phenotype
MIM: 612499 MIM: 610889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIMT1 PubMed Links

Gene Details

Gene
DIMT1
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
There are no transcripts associated with this gene.

Gene
IPO11
Gene Name
importin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134779.1 Intron NP_001128251.1
NM_016338.4 Intron NP_057422.3

View Full Product Details