Product Details

SNP ID
rs6963359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:49778650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATGCTTTTTTTGAAGAATGTTG[A/G]AAATATGAATGTAGAATGGTATAGG
Phenotype
MIM: 611108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VWC2 PubMed Links

Gene Details

Gene
VWC2
Gene Name
von Willebrand factor C domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198570.4 Intron NP_940972.2

View Full Product Details