Product Details

SNP ID
rs6665033
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:226871273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGTTTTATCAGGCATTTCCAGC[A/G]GTGAGGAGACAGCCAGAAGCAAGCT
Phenotype
MIM: 600759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSEN2 PubMed Links

Gene Details

Gene
PSEN2
Gene Name
presenilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000447.2 411 UTR 5 NP_000438.2
NM_012486.2 411 UTR 5 NP_036618.2
XM_005273199.3 411 Intron XP_005273256.1
XM_017001835.1 411 UTR 5 XP_016857324.1
XM_017001836.1 411 Intron XP_016857325.1

View Full Product Details