Product Details

SNP ID
rs7060633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24561228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCAGAGGAGTATTATGGTGGAG[C/T]GCAGGAGTGGGGTCTATTATAAAAT
Phenotype
MIM: 300948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCYT1B PubMed Links

Gene Details

Gene
PCYT1B
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163264.1 2233 UTR 3 NP_001156736.1
NM_001163265.1 2233 UTR 3 NP_001156737.1
NM_004845.4 2233 UTR 3 NP_004836.2
XM_017029977.1 2233 UTR 3 XP_016885466.1

View Full Product Details