Product Details

SNP ID
rs7972639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64192648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGTAACAACTCAATTACTCTTT[C/G]TGGCTTCTTTTTATTTTGAATCAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf66 PubMed Links

Gene Details

Gene
C12orf66
Gene Name
chromosome 12 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300940.1 2432 Intron NP_001287869.1
NM_001300941.1 2432 UTR 3 NP_001287870.1
NM_152440.4 2432 UTR 3 NP_689653.3

View Full Product Details