Product Details

SNP ID
rs8103557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17215101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGAGGAAAAGCAAGTGATTCGCC[G/T]AAGTAAGAATGGGGTGGCGCCTCTC
Phenotype
MIM: 602129 MIM: 610675
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYO9B PubMed Links

Gene Details

Gene
MYO9B
Gene Name
myosin IXB
There are no transcripts associated with this gene.

Gene
USE1
Gene Name
unconventional SNARE in the ER 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018467.3 466 Intron NP_060937.1
XM_017026975.1 466 UTR 5 XP_016882464.1
XM_017026976.1 466 Intron XP_016882465.1

View Full Product Details