Product Details

SNP ID
rs4387594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69142443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCAGACAACGTCTCCAGTATT[A/C]TGCTGACGTGCATTTTGGGAGCTTC
Phenotype
MIM: 613202 MIM: 607456
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHTF8 PubMed Links

Gene Details

Gene
CHTF8
Gene Name
chromosome transmission fidelity factor 8
There are no transcripts associated with this gene.

Gene
UTP4
Gene Name
UTP4, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318391.1 Intron NP_001305320.1
NM_032830.2 Intron NP_116219.2

View Full Product Details