Product Details
- SNP ID
-
rs6962301
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:155648503 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCATTCGAAATTGTGAAAGTGAAA[C/T]AAGTTTTAGTACTTCAACAGGTAAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100506302
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115196199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100506302
- Gene Name
- uncharacterized LOC100506302
There are no transcripts associated with this gene.
- Gene
- RBM33
- Gene Name
- RNA binding motif protein 33
View Full Product Details