Product Details
- SNP ID
-
rs7909557
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:7161054 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACAGTGCTCACCCATGGTGACC[C/G]CAACTGCACAGAGAAACGCTGAAGT
- Phenotype
-
MIM: 615392
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SFMBT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114503012] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SFMBT2
- Gene Name
- Scm-like with four mbt domains 2
View Full Product Details