Product Details
- SNP ID
-
rs7639979
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46733286 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTCCAGGAATTTGGGGGTACCTC[A/G]GGATAACCAGTACACAAGATGGATA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS45
PubMed Links
Gene Details
- Gene
- PRSS45
- Gene Name
- protease, serine 45
There are no transcripts associated with this gene.
- Gene
- PRSS46
- Gene Name
- protease, serine 46
View Full Product Details