Product Details

SNP ID
rs7108947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17494636 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGAGGAACAGGGGCAAGAGTCC[C/T]TTGCAAAAGGCAAGTAACAGAAAAC
Phenotype
MIM: 605242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USH1C PubMed Links

Gene Details

Gene
USH1C
Gene Name
USH1 protein network component harmonin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297764.1 3697 Intron NP_001284693.1
NM_005709.3 3697 Intron NP_005700.2
NM_153676.3 3697 Intron NP_710142.1
XM_011519832.2 3697 Intron XP_011518134.1
XM_011519834.2 3697 Intron XP_011518136.1
XM_017017072.1 3697 Intron XP_016872561.1
XM_017017073.1 3697 Intron XP_016872562.1
XM_017017074.1 3697 Intron XP_016872563.1
XM_017017075.1 3697 UTR 3 XP_016872564.1

View Full Product Details