Product Details

SNP ID
rs6463240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44623274 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCATCCATGTCCTTTTGTCTT[C/G]AGGCACACCCACTTGGAAGGAGCCC
Phenotype
MIM: 613022
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OGDH PubMed Links

Gene Details

Gene
OGDH
Gene Name
oxoglutarate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003941.2 Intron NP_001003941.1
NM_001165036.1 Intron NP_001158508.1
NM_002541.3 Intron NP_002532.2
XM_005249759.4 Intron XP_005249816.1
XM_005249761.3 Intron XP_005249818.1
XM_011515408.2 Intron XP_011513710.1

View Full Product Details