Product Details

SNP ID
rs6864662
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141121852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGTTACAAGCAGTGCAGGTTTA[C/T]CAACGGCTTGGGGCAGCGATATACT
Phenotype
MIM: 606330
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PCDHB4 PubMed Links

Gene Details

Gene
PCDHB4
Gene Name
protocadherin beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018938.3 103 UTR 5 NP_061761.1

View Full Product Details