Product Details
- SNP ID
-
rs7849494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:86033068 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTGAATATGTTTCTAGAAACGAT[A/G]CAGACTTTCCGAATGGTTTTCATCA
- Phenotype
-
MIM: 606804
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GOLM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139971378] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GOLM1
- Gene Name
- golgi membrane protein 1
- Gene
- NAA35
- Gene Name
- N(alpha)-acetyltransferase 35, NatC auxiliary subunit
There are no transcripts associated with this gene.
View Full Product Details