Product Details

SNP ID
rs8012929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102375646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCATAAATCCATGATGCAAGAAG[G/T]TTCTGTGAGAGGAGCTTGGAAGTCT
Phenotype
MIM: 615000
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TECPR2 PubMed Links

Gene Details

Gene
TECPR2
Gene Name
tectonin beta-propeller repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172631.2 Intron NP_001166102.1
NM_014844.4 Intron NP_055659.2

View Full Product Details