Product Details

SNP ID
rs7310043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21765468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTTCAAATTGAGAGAAACGAGC[A/T]TACCCACCCCTGCACATAACTTAAG
Phenotype
MIM: 600935
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ8 PubMed Links

Gene Details

Gene
KCNJ8
Gene Name
potassium voltage-gated channel subfamily J member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004982.3 1845 UTR 3 NP_004973.1
XM_005253358.4 1845 UTR 3 XP_005253415.1
XM_017019283.1 1845 UTR 3 XP_016874772.1
XM_017019284.1 1845 UTR 3 XP_016874773.1

View Full Product Details