Product Details

SNP ID
rs7497610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78882530 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTGACTCCTAGTGTATCTCATA[G/T]CTTTTTATTACTAAACAATGCCTTG
Phenotype
MIM: 607303
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MORF4L1 PubMed Links

Gene Details

Gene
MORF4L1
Gene Name
mortality factor 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265603.1 Intron NP_001252532.1
NM_001265604.1 Intron NP_001252533.1
NM_001265605.1 Intron NP_001252534.1
NM_006791.3 Intron NP_006782.1
NM_206839.2 Intron NP_996670.1
XM_017021872.1 Intron XP_016877361.1

View Full Product Details