Product Details

SNP ID
rs6438505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119217225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATCACCCCAGTGAAGTACATTT[A/G]GGGAAGATAATGGGGATTGTTTATC
Phenotype
MIM: 604015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALT4 PubMed Links

Gene Details

Gene
B4GALT4
Gene Name
beta-1,4-galactosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003778.3 Intron NP_003769.1
NM_212543.1 Intron NP_997708.1
XM_005247855.1 Intron XP_005247912.1
XM_006713798.2 Intron XP_006713861.1
XM_006713799.2 Intron XP_006713862.1
XM_006713800.1 Intron XP_006713863.1
XM_006713801.2 Intron XP_006713864.1
XM_011513260.1 Intron XP_011511562.1
XM_017007401.1 Intron XP_016862890.1
Gene
B4GALT4-AS1
Gene Name
B4GALT4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details