Product Details

SNP ID
rs7305779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55130076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTTTTCTATTCATCTGTTATTG[A/C]ACCCAAGGCTATGATCAACTTCTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR9K2 PubMed Links

Gene Details

Gene
OR9K2
Gene Name
olfactory receptor family 9 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005243.1 308 Missense Mutation GAA,GCA E103A NP_001005243.1

View Full Product Details