Product Details

SNP ID
rs7909811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97641009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGCTCTGGCCGCTCAGGCCGC[A/G]GCGGCAGCCCACGCCGCTCAGGCCC
Phenotype
MIM: 609763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MORN4 PubMed Links

Gene Details

Gene
MORN4
Gene Name
MORN repeat containing 4
There are no transcripts associated with this gene.

Gene
PI4K2A
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018425.3 324 Silent Mutation GCA,GCG A89A NP_060895.1

View Full Product Details