Product Details

SNP ID
rs7078759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97587011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTAACCTTTCAAATCCAGTCCTC[A/G]TTTTAGAATAGGAACTGAGCCTTCA
Phenotype
MIM: 610734 MIM: 613597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD2 PubMed Links

Gene Details

Gene
ANKRD2
Gene Name
ankyrin repeat domain 2
There are no transcripts associated with this gene.

Gene
C10orf62
Gene Name
chromosome 10 open reading frame 62
There are no transcripts associated with this gene.

Gene
HOGA1
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134670.1 Intron NP_001128142.1
NM_138413.3 Intron NP_612422.2

View Full Product Details