Product Details

SNP ID
rs6950177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103100236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAAAAGGCACATTTCTTTATAAC[A/G]GTTTTTTCAAATCTACCTCCCTATT
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161009.2 4703 Intron NP_001154481.1
NM_001161010.2 4703 Intron NP_001154482.1
NM_001161011.2 4703 Intron NP_001154483.1
NM_001161012.2 4703 Intron NP_001154484.1
NM_001161013.2 4703 Intron NP_001154485.1
NM_031905.4 4703 Intron NP_114111.2
XM_005250628.2 4703 Intron XP_005250685.1
XM_011516601.2 4703 Intron XP_011514903.1
XM_011516602.2 4703 Intron XP_011514904.1
XM_017012684.1 4703 Intron XP_016868173.1
XM_017012685.1 4703 Intron XP_016868174.1
XM_017012686.1 4703 Intron XP_016868175.1
XM_017012687.1 4703 Intron XP_016868176.1
XM_017012688.1 4703 Intron XP_016868177.1
XM_017012689.1 4703 Intron XP_016868178.1
Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 4703 UTR 3 NP_001116310.1
NM_198990.4 4703 UTR 3 NP_945341.3
XM_005250214.4 4703 UTR 3 XP_005250271.2
XM_005250215.2 4703 UTR 3 XP_005250272.1
XM_005250218.2 4703 UTR 3 XP_005250275.1
XM_011515933.2 4703 UTR 3 XP_011514235.1
XM_011515934.2 4703 UTR 3 XP_011514236.1
XM_017011854.1 4703 UTR 3 XP_016867343.1
XM_017011855.1 4703 UTR 3 XP_016867344.1
XM_017011856.1 4703 Intron XP_016867345.1
XM_017011857.1 4703 Intron XP_016867346.1

View Full Product Details