Product Details

SNP ID
rs6955219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123569486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGATCCTATGTGGCCTTAAAATG[C/T]TATTGCATGCATTTTTAATAGTTAG
Phenotype
MIM: 601677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB15 PubMed Links

Gene Details

Gene
ASB15
Gene Name
ankyrin repeat and SOCS box containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290258.1 Intron NP_001277187.1
NM_080928.4 Intron NP_563616.3
XM_005250146.4 Intron XP_005250203.1
XM_011515816.2 Intron XP_011514118.1
XM_011515817.2 Intron XP_011514119.1
XM_011515818.2 Intron XP_011514120.1
XM_011515819.2 Intron XP_011514121.1
XM_011515820.2 Intron XP_011514122.1
XM_011515821.2 Intron XP_011514123.1
XM_017011756.1 Intron XP_016867245.1
XM_017011757.1 Intron XP_016867246.1
XM_017011758.1 Intron XP_016867247.1
Gene
NDUFA5
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282419.2 Intron NP_001269348.1
NM_001282420.2 Intron NP_001269349.1
NM_001282421.2 Intron NP_001269350.1
NM_001282422.2 Intron NP_001269351.1
NM_001291304.1 Intron NP_001278233.1
NM_005000.4 Intron NP_004991.1

View Full Product Details