Product Details

SNP ID
rs6914852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30184659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAATGGTGGTCAAGTGACCAC[A/T]GAGGCCCAGAGCCGTTGGAACAGTC
Phenotype
MIM: 600830
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM26 PubMed Links

Gene Details

Gene
TRIM26
Gene Name
tripartite motif containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242783.1 2967 UTR 3 NP_001229712.1
NM_003449.4 2967 UTR 3 NP_003440.1
XM_005249374.2 2967 UTR 3 XP_005249431.1
XM_005249375.2 2967 UTR 3 XP_005249432.1
XM_005249376.2 2967 UTR 3 XP_005249433.1
XM_005249377.2 2967 UTR 3 XP_005249434.1
XM_005249378.2 2967 UTR 3 XP_005249435.1
XM_006715180.2 2967 UTR 3 XP_006715243.1
XM_011514859.1 2967 UTR 3 XP_011513161.1
XM_017011263.1 2967 UTR 3 XP_016866752.1

View Full Product Details