Product Details

SNP ID
rs6964700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:79457001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGCACTCTGAGTGTAGAGCAGA[A/G]TGCTAAATATTGCATGCTATTCCAG
Phenotype
MIM: 606382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGI2 PubMed Links

Gene Details

Gene
MAGI2
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
There are no transcripts associated with this gene.

Gene
MAGI2-AS3
Gene Name
MAGI2 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details