Product Details

SNP ID
rs6426490
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:228014306 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGAAAGAGAGAAAGAGCAAACAA[A/G]CAAGCGAGTAACAGGGCCCGAGTTG
Phenotype
MIM: 606359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT3A PubMed Links

Gene Details

Gene
WNT3A
Gene Name
Wnt family member 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033131.3 Intron NP_149122.1
XM_011544319.2 Intron XP_011542621.1

View Full Product Details