Product Details

SNP ID
rs7258368
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12146097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTATTAAGGGATGCATGACCCA[C/T]GCTGACTTCTCCACATGATTTTACT
Phenotype
MIM: 194557
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF20 PubMed Links

Gene Details

Gene
ZNF20
Gene Name
zinc finger protein 20
There are no transcripts associated with this gene.

Gene
ZNF625
Gene Name
zinc finger protein 625
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145233.3 492 Missense Mutation ATG,GTG M107V NP_660276.2
Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details