Product Details
- SNP ID
-
hCV29183587
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:100630456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTCTCTCCAGGCCAAAACATGT[A/T]AAAAAAGGGACCACTTATGATCTTT
- Phenotype
-
MIM: 300191
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN6
PubMed Links
Gene Details
- Gene
- TSPAN6
- Gene Name
- tetraspanin 6
There are no transcripts associated with this gene.
View Full Product Details