Product Details
- SNP ID
-
rs7877907
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:115565492 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCATTGACTCATAATTTGACCTT[C/T]GGAAAGTCATTTAAAATTATTATAT
- Phenotype
-
MIM: 300131
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLS3
PubMed Links
Gene Details
- Gene
- PLS3
- Gene Name
- plastin 3
- Gene
- PLS3-AS1
- Gene Name
- PLS3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details