Product Details
- SNP ID
-
rs7251816
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:50798430 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGTGATGGCCTGGATCTCCAAGA[C/T]GGCCTCCAGCACTGTCATGCCCTTG
- Phenotype
-
MIM: 606362
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACPT
PubMed Links
Gene Details
- Gene
- ACPT
- Gene Name
- acid phosphatase, testicular
There are no transcripts associated with this gene.
- Gene
- C19orf48
- Gene Name
- chromosome 19 open reading frame 48
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290149.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277078.1 |
NM_001290150.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277079.1 |
NM_001290151.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277080.1 |
NM_001290152.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277081.1 |
NM_001290153.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277082.1 |
NM_001290154.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277083.1 |
NM_001290155.1 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_001277084.1 |
NM_032712.2 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_116101.1 |
NM_199249.2 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_954857.1 |
NM_199250.2 |
905 |
Missense Mutation |
ATC,GTC |
I7V |
NP_954858.1 |
- Gene
- SNORD88A
- Gene Name
- small nucleolar RNA, C/D box 88A
There are no transcripts associated with this gene.
- Gene
- SNORD88B
- Gene Name
- small nucleolar RNA, C/D box 88B
There are no transcripts associated with this gene.
- Gene
- SNORD88C
- Gene Name
- small nucleolar RNA, C/D box 88C
There are no transcripts associated with this gene.
View Full Product Details