Product Details
- SNP ID
-
rs7165874
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:60492410 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTAGCTCTTGTAATGCATCAGAT[A/T]TAAACTACACATGTAGAAGTAAGAA
- Phenotype
-
MIM: 600825
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RORA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114653976] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RORA
- Gene Name
- RAR related orphan receptor A
- Gene
- RORA-AS1
- Gene Name
- RORA antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details