Product Details

SNP ID
rs6440429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146396176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTATGTATACACACGTATACACTT[A/G]ATTCATATTATTTGCTTTAATTTAA
Phenotype
MIM: 607610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLSCR2 PubMed Links

Gene Details

Gene
PLSCR2
Gene Name
phospholipid scramblase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199978.1 Intron NP_001186907.1
NM_001199979.1 Intron NP_001186908.1
NM_020359.2 Intron NP_065092.1
XM_011513013.2 Intron XP_011511315.2
XM_011513019.2 Intron XP_011511321.2
XM_011513020.2 Intron XP_011511322.2
XM_011513021.2 Intron XP_011511323.2
XM_011513022.2 Intron XP_011511324.2
XM_011513023.2 Intron XP_011511325.2
XM_017006898.1 Intron XP_016862387.1
XM_017006899.1 Intron XP_016862388.1
XM_017006900.1 Intron XP_016862389.1
XM_017006901.1 Intron XP_016862390.1
XM_017006902.1 Intron XP_016862391.1
XM_017006903.1 Intron XP_016862392.1
XM_017006904.1 Intron XP_016862393.1
XM_017006905.1 Intron XP_016862394.1
XM_017006906.1 Intron XP_016862395.1
XM_017006907.1 Intron XP_016862396.1
XM_017006908.1 Intron XP_016862397.1
XM_017006909.1 Intron XP_016862398.1
XM_017006910.1 Intron XP_016862399.1
XM_017006911.1 Intron XP_016862400.1
XM_017006912.1 Intron XP_016862401.1
XM_017006913.1 Intron XP_016862402.1
XM_017006914.1 Intron XP_016862403.1
XM_017006915.1 Intron XP_016862404.1

View Full Product Details