Product Details
- SNP ID
-
rs6898962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:33988130 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGATGGCACCCGGATTAGATTGT[A/G]GAATCTACCCCTTCCTCACATGCCT
- Phenotype
-
MIM: 604489
MIM: 606202
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMACR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150664086] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMACR
- Gene Name
- alpha-methylacyl-CoA racemase
- Gene
- C1QTNF3-AMACR
- Gene Name
- C1QTNF3-AMACR readthrough (NMD candidate)
There are no transcripts associated with this gene.
- Gene
- SLC45A2
- Gene Name
- solute carrier family 45 member 2
There are no transcripts associated with this gene.
View Full Product Details