Product Details

SNP ID
rs7597878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178474589 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGAAATGAAAGCTCATAGAGGA[C/T]TGGGAGTAGTGTCTACTTTGGTCAC
Phenotype
MIM: 607062 MIM: 607774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP7 PubMed Links

Gene Details

Gene
FKBP7
Gene Name
FK506 binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135212.1 Intron NP_001128684.1
NM_181342.2 Intron NP_851939.1
XM_005246638.4 Intron XP_005246695.1
XM_011511348.2 Intron XP_011509650.1
XM_011511349.2 Intron XP_011509651.1
Gene
PLEKHA3
Gene Name
pleckstrin homology domain containing A3
There are no transcripts associated with this gene.

View Full Product Details