Product Details

SNP ID
rs8067658
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29049533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCAATCTGCCCATGGAAGCCATC[C/T]ATCTTCATACTGGGAGGAAAAGGCC
Phenotype
MIM: 616713
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PIPOX PubMed Links

Gene Details

Gene
PIPOX
Gene Name
pipecolic acid and sarcosine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016518.2 Intron NP_057602.2
XM_017024722.1 Intron XP_016880211.1

View Full Product Details