Product Details

SNP ID
rs7706625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34998151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATATCAAGAGGCCCTGAGGTAG[A/G]GTGGCTCCAGGAATGCTTAAGTGAG
Phenotype
MIM: 612471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGXT2 PubMed Links

Gene Details

Gene
AGXT2
Gene Name
alanine--glyoxylate aminotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306173.1 2406 UTR 3 NP_001293102.1
NM_031900.3 2406 UTR 3 NP_114106.1
XM_005248337.2 2406 UTR 3 XP_005248394.1
XM_005248338.2 2406 UTR 3 XP_005248395.1
XM_017009748.1 2406 UTR 3 XP_016865237.1

View Full Product Details