Product Details

SNP ID
rs6668667
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:1179288 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGGACCCGAGCCGGCTTCAAGAGG[A/G]GCAAGAGGCCAAGGATCCAGCAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTLL10 PubMed Links

Gene Details

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130045.1 383 Missense Mutation AGC,GGC S25G NP_001123517.1
NM_153254.2 383 Intron NP_694986.2
XM_005244738.1 383 Intron XP_005244795.1
XM_011541177.2 383 Missense Mutation AGC,GGC S25G XP_011539479.1
XM_017000906.1 383 Missense Mutation AGC,GGC S25G XP_016856395.1
XM_017000907.1 383 Missense Mutation AGC,GGC S25G XP_016856396.1
XM_017000908.1 383 Missense Mutation AGC,GGC S25G XP_016856397.1
XM_017000909.1 383 Missense Mutation AGC,GGC S25G XP_016856398.1
XM_017000910.1 383 Missense Mutation AGC,GGC S25G XP_016856399.1
XM_017000911.1 383 Missense Mutation AGC,GGC S25G XP_016856400.1
XM_017000912.1 383 Missense Mutation AGC,GGC S25G XP_016856401.1
Gene
TTLL10-AS1
Gene Name
TTLL10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details