Product Details

SNP ID
rs7952602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126363774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTTCTCCCAGGGGAAGCTGCTC[C/G]AATGAGGGGAGTTGGCCAACACAGT
Phenotype
MIM: 104240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ST3GAL4 PubMed Links

Gene Details

Gene
ST3GAL4
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254757.1 Intron NP_001241686.1
NM_001254758.1 Intron NP_001241687.1
NM_001254759.1 Intron NP_001241688.1
NM_006278.2 Intron NP_006269.1
XM_011542959.1 Intron XP_011541261.1
XM_011542960.2 Intron XP_011541262.1
XM_017018134.1 Intron XP_016873623.1
Gene
ST3GAL4-AS1
Gene Name
ST3GAL4 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details