Product Details

SNP ID
rs7129774
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74180443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATAAGAACAACGTTATTTCCCCA[C/T]CAGTTCCCTTAAGATGGTATAACTA
Phenotype
MIM: 615944 MIM: 611117
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C2CD3 PubMed Links
Additional Information
For this assay, SNP(s) [rs80163117] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C2CD3
Gene Name
C2 calcium dependent domain containing 3
There are no transcripts associated with this gene.

Gene
PPME1
Gene Name
protein phosphatase methylesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271593.1 Intron NP_001258522.1
NM_016147.2 Intron NP_057231.1
XM_017017913.1 Intron XP_016873402.1

View Full Product Details