Product Details

SNP ID
rs7263778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:51390575 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAGGTGCTTGCTCTCTGCAGC[C/T]TGGGTCAGCCCCCCAAGTCTATGGA
Phenotype
MIM: 600490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFATC2 PubMed Links

Gene Details

Gene
NFATC2
Gene Name
nuclear factor of activated T-cells 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136021.2 3120 UTR 3 NP_001129493.1
NM_001258292.1 3120 UTR 3 NP_001245221.1
NM_001258294.1 3120 UTR 3 NP_001245223.1
NM_001258295.1 3120 UTR 3 NP_001245224.1
NM_001258296.1 3120 UTR 3 NP_001245225.1
NM_001258297.1 3120 UTR 3 NP_001245226.1
NM_012340.4 3120 UTR 3 NP_036472.2
NM_173091.3 3120 UTR 3 NP_775114.1
XM_011528824.2 3120 Intron XP_011527126.1
XM_011528825.2 3120 Intron XP_011527127.1
XM_011528826.2 3120 Intron XP_011527128.1
XM_017027850.1 3120 Intron XP_016883339.1
XM_017027851.1 3120 UTR 3 XP_016883340.1

View Full Product Details