Product Details

SNP ID
rs7146660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105323060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGCCGTGACAGGCTGGAGGGAGA[C/T]GTCTGGGTGGCTTTGTGGTCATGTC
Phenotype
MIM: 604902 MIM: 610423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRF1 PubMed Links

Gene Details

Gene
BRF1
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
There are no transcripts associated with this gene.

Gene
PACS2
Gene Name
phosphofurin acidic cluster sorting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100913.2 Intron NP_001094383.2
NM_001243127.2 Intron NP_001230056.1
NM_015197.3 Intron NP_056012.2
XM_006720090.1 Intron XP_006720153.1
XM_006720091.1 Intron XP_006720154.1
XM_006720092.2 Intron XP_006720155.1
XM_017021105.1 Intron XP_016876594.1
XM_017021106.1 Intron XP_016876595.1
XM_017021107.1 Intron XP_016876596.1
XM_017021108.1 Intron XP_016876597.1
XM_017021109.1 Intron XP_016876598.1
XM_017021110.1 Intron XP_016876599.1
XM_017021111.1 Intron XP_016876600.1
XM_017021112.1 Intron XP_016876601.1

View Full Product Details