Product Details

SNP ID
rs7079328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43554297 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCAAAGAGTTAGGTTTTGGTTC[C/T]ATTAAATTTTCTCTATTGTTTGTCC
Phenotype
MIM: 601069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF239 PubMed Links

Gene Details

Gene
ZNF239
Gene Name
zinc finger protein 239
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099282.1 Intron NP_001092752.1
NM_001099283.1 Intron NP_001092753.1
NM_001099284.1 Intron NP_001092754.1
NM_001324347.1 Intron NP_001311276.1
NM_001324348.1 Intron NP_001311277.1
NM_001324349.1 Intron NP_001311278.1
NM_001324350.1 Intron NP_001311279.1
NM_001324351.1 Intron NP_001311280.1
NM_001324352.1 Intron NP_001311281.1
NM_001324353.1 Intron NP_001311282.1
NM_005674.2 Intron NP_005665.2
XM_005271832.2 Intron XP_005271889.1
XM_006718001.2 Intron XP_006718064.1
XM_006718003.3 Intron XP_006718066.1
XM_011540232.2 Intron XP_011538534.1
XM_011540234.2 Intron XP_011538536.1
XM_011540235.2 Intron XP_011538537.1
XM_011540236.2 Intron XP_011538538.1
XM_011540237.2 Intron XP_011538539.1
XM_011540238.2 Intron XP_011538540.1
XM_017016740.1 Intron XP_016872229.1

View Full Product Details