Product Details
- SNP ID
-
rs6565543
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81179401 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCAGGTGCCTGGTCCTGCGGGG[A/C]GCACATGTGTGCACGTGTGTGCTGG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AATK-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143461333] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AATK-AS1
- Gene Name
- AATK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details