Product Details

SNP ID
rs6502774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1280060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACAGGAGCCAGGCTCCGCCGCAT[C/T]CCTGGACCTGCCGGAGATGGAGATA
Phenotype
MIM: 615416 MIM: 612211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BHLHA9 PubMed Links

Gene Details

Gene
BHLHA9
Gene Name
basic helix-loop-helix family member a9
There are no transcripts associated with this gene.

Gene
TUSC5
Gene Name
tumor suppressor candidate 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172367.2 398 Missense Mutation TCC,TTC S20F NP_758955.2

View Full Product Details