Product Details

SNP ID
rs8071342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58242878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCTTTCCTGTTCCAATATTAG[A/G]GTGTCTGGTGCTCCTTTGGCTTTCT
Phenotype
MIM: 150205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPO PubMed Links

Gene Details

Gene
LPO
Gene Name
lactoperoxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160102.1 Intron NP_001153574.1
NM_006151.2 Intron NP_006142.1
XM_011524808.2 Intron XP_011523110.1
XM_011524810.2 Intron XP_011523112.1

View Full Product Details