Product Details
- SNP ID
-
rs8053782
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:87696368 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCCAGGACCAGCAACCTCAGAC[G/T]TACAGGCAGCTGGGGCTTCTCTCCC
- Phenotype
-
MIM: 605268
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
JPH3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116409000] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JPH3
- Gene Name
- junctophilin 3
- Gene
- KLHDC4
- Gene Name
- kelch domain containing 4
- Gene
- LOC100129215
- Gene Name
- uncharacterized LOC100129215
There are no transcripts associated with this gene.
- Gene
- LOC105371397
- Gene Name
- uncharacterized LOC105371397
There are no transcripts associated with this gene.
View Full Product Details